Assay Details
Gene Symbol: Arhgef18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133962.3 | NP_598723.3 | 1 - 2 | 99 | 77 | |
GenBank mRNA | AK041423.1 | - | 1 - 2 | 102 | 77 | |
AK147923.1 | - | 1 - 2 | 70 | 77 | ||
BC060639.1 | - | 1 - 2 | 71 | 6849875 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK041423.1, BC060639.1, NM_133962.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
