Assay Details
Gene Symbol: Arhgef18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133962.3 | NP_598723.3 | 15 - 16 | 2098 | 74 | |
GenBank mRNA | AK147923.1 | - | 15 - 16 | 2071 | 74 | |
AK172963.1 | - | 9 - 10 | 1201 | 74 | ||
BC034512.1 | - | 4 - 5 | 357 | 74 | ||
BC060639.1 | - | 15 - 16 | 2070 | 6849875 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
