Assay Details
Gene Symbol: Poln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289803.1 | NP_001276732.1 | 15 - 16 | 1881 | 84 | |
NM_001289804.1 | NP_001276733.1 | 17 - 18 | 1984 | 84 | ||
NM_181857.4 | NP_862905.1 | 16 - 17 | 1935 | 84 | ||
GenBank mRNA | AK133063.1 | - | 17 - 18 | 1904 | 84 | |
AY135562.3 | - | 16 - 17 | 1857 | 84 | ||
BC141101.1 | - | 16 - 17 | 1835 | 9055713 | 84 | |
BC145378.1 | - | 15 - 16 | 1781 | 9053936 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
