Assay Details
Gene Symbol: Gabrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008069.4 | NP_032095.1 | 5 - 6 | 551 | 79 | |
GenBank mRNA | BC130258.1 | - | 5 - 6 | 555 | 40142758 | 79 |
BC157920.1 | - | 5 - 6 | 617 | 9007350 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
