Assay Details
Gene Symbol: Gckr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144909.1 | NP_659158.1 | 15 - 16 | 1376 | 93 | |
GenBank mRNA | AK149377.1 | - | 13 - 14 | 3275 | 93 | |
BC012412.1 | - | 15 - 16 | 1376 | 4159892 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
