Assay Details
Gene Symbol: Epha5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007937.3 | NP_031963.2 | 11 - 12 | 2696 | 102 | |
GenBank mRNA | BC057401.1 | - | 10 - 11 | 2605 | 5697758 | 102 |
U07357.1 | - | 11 - 12 | 2236 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
