Assay Details
Gene Symbol: Epha5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007937.3 | NP_031963.2 | 9 - 10 | 2442 | 142 | |
GenBank mRNA | BC057401.1 | - | 8 - 9 | 2351 | 5697758 | 142 |
BU707304.1 | - | 5 - 6 | 462 | 141 | ||
U07357.1 | - | 9 - 10 | 1982 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
