Assay Details
Gene Symbol: Champ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181854.2 | NP_862902.1 | 1 - 2 | 167 | 71 | |
GenBank mRNA | AK129444.1 | - | 1 - 2 | 67 | 71 | |
AK132164.1 | - | 1 - 2 | 130 | 71 | ||
AK133990.1 | - | 1 - 2 | 93 | 71 | ||
AK170835.1 | - | 1 - 2 | 110 | 71 | ||
BC028991.1 | - | 1 - 2 | 79 | 5355869 | 71 | |
CJ077618.1 | - | 1 - 2 | 168 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK132164.1, AK133990.1, AK170835.1, BC028991.1, NM_181854.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
