Assay Details
Gene Symbol: Cox7b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030052.3 | NP_084328.1 | 2 - 3 | 94 | 112 | |
GenBank mRNA | AK019665.1 | - | 2 - 3 | 95 | 112 | |
AK076833.1 | - | 2 - 3 | 95 | 112 | ||
BC059709.1 | - | 2 - 3 | 89 | 6741029 | 112 | |
BY099104.1 | - | 2 - 3 | 94 | 112 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK019665.1, AK076833.1, BC059709.1, NM_030052.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
