Assay Details
Gene Symbol: Dcp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133761.3 | NP_598522.3 | 8 - 9 | 1567 | 77 | |
GenBank mRNA | AJ344447.1 | - | 8 - 9 | 1567 | 77 | |
AK016254.1 | - | 6 - 7 | 1246 | 77 | ||
BC066173.1 | - | 8 - 9 | 1520 | 30049516 | 77 | |
CK392647.1 | - | 2 - 3 | 481 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
