Assay Details
Gene Symbol: Cdh17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019753.4 | NP_062727.1 | 4 - 5 | 425 | 74 | |
GenBank mRNA | AF177669.1 | - | 4 - 5 | 321 | 74 | |
AK089791.1 | - | 4 - 5 | 392 | 74 | ||
AK155633.1 | - | 4 - 5 | 426 | 74 | ||
AK171733.1 | - | 4 - 5 | 348 | 74 | ||
BC042891.1 | - | 4 - 5 | 343 | 4208360 | 74 | |
D87912.1 | - | 4 - 5 | 370 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
