Assay Details
Gene Symbol: Mark2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080388.2 | NP_001073857.1 | 6 - 7 | 1110 | 92 | |
NM_001080389.2 | NP_001073858.1 | 6 - 7 | 1110 | 92 | ||
NM_001080390.2 | NP_001073859.1 | 6 - 7 | 827 | 92 | ||
NM_007928.3 | NP_031954.2 | 6 - 7 | 1110 | 92 | ||
GenBank mRNA | AK045329.1 | - | 6 - 7 | 684 | 92 | |
AK172444.1 | - | 6 - 7 | 1070 | 92 | ||
AK172664.1 | - | 6 - 7 | 753 | 92 | ||
AK215655.1 | - | 2 - 3 | 121 | 92 | ||
AK217185.1 | - | 1 - 2 | 70 | 92 | ||
AK220191.1 | - | 7 - 8 | 676 | 92 | ||
BC058556.1 | - | 6 - 7 | 1047 | 5289799 | 92 | |
X70764.1 | - | 6 - 7 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
