Assay Details
Gene Symbol: Gria2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039195.1 | NP_001034284.1 | 2 - 3 | 637 | 62 | |
NM_001083806.1 | NP_001077275.1 | 2 - 3 | 637 | 62 | ||
NM_013540.2 | NP_038568.2 | 2 - 3 | 637 | 62 | ||
GenBank mRNA | AB109210.1 | - | 2 - 3 | 230 | 62 | |
AB109211.1 | - | 2 - 3 | 230 | 62 | ||
AB111957.1 | - | 2 - 3 | 230 | 62 | ||
AK014389.1 | - | 2 - 3 | 682 | 62 | ||
AK043490.1 | - | 2 - 3 | 610 | 62 | ||
AK044574.1 | - | 2 - 3 | 569 | 62 | ||
AK046861.1 | - | 2 - 3 | 637 | 62 | ||
AK134572.1 | - | 2 - 3 | 568 | 62 | ||
AK134647.1 | - | 1 - 2 | 83 | 62 | ||
AK134772.1 | - | 1 - 2 | 208 | 62 | ||
AK141525.1 | - | 2 - 3 | 612 | 62 | ||
BC048248.1 | - | 2 - 3 | 496 | 6494013 | 62 | |
BC066193.1 | - | 2 - 3 | 425 | 62 | ||
X57498.1 | - | 2 - 3 | 432 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
