Assay Details
Gene Symbol: Prpf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133701.2 | NP_598462.1 | 1 - 2 | 214 | 72 | |
GenBank mRNA | AK011639.1 | - | 1 - 2 | 156 | 72 | |
AK081998.1 | - | 1 - 2 | 149 | 72 | ||
AK145461.1 | - | 1 - 2 | 217 | 72 | ||
AK166890.1 | - | 1 - 2 | 166 | 72 | ||
BC014869.1 | - | 1 - 2 | 141 | 3602158 | 72 | |
BC023691.1 | - | 1 - 2 | 143 | 5344332 | 72 | |
BC025030.1 | - | 1 - 2 | 160 | 4947416 | 72 | |
BY735790.1 | - | 1 - 2 | 214 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
