Assay Details
Gene Symbol: Grtp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025768.2 | NP_080044.2 | 1 - 2 | 140 | 78 | |
GenBank mRNA | AK007905.1 | - | 1 - 2 | 145 | 78 | |
AK017197.1 | - | 1 - 2 | 104 | 78 | ||
AK017251.1 | - | 1 - 2 | 51 | 78 | ||
AK044437.1 | - | 104 | 78 | |||
AK082863.1 | - | 1 - 2 | 134 | 78 | ||
AK149322.1 | - | 137 | 78 | |||
BC029096.1 | - | 1 - 2 | 146 | 3500563 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK007905.1, AK017197.1, AK017251.1, AK082863.1, AK149322.1, BC029096.1, NM_025768.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
