Assay Details
Gene Symbol: Chat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009891.2 | NP_034021.1 | 5 - 6 | 712 | 67 | |
GenBank mRNA | AK046406.1 | - | 5 - 6 | 713 | 67 | |
AK047708.1 | - | 4 - 5 | 604 | 67 | ||
AK143147.1 | - | 4 - 5 | 610 | 67 | ||
BC119322.1 | - | 4 - 5 | 474 | 8734071 | 67 | |
BC120589.1 | - | 4 - 5 | 474 | 40129512 | 67 | |
D12487.1 | - | 4 - 5 | 636 | 67 | ||
D12490.1 | - | 4 - 5 | 605 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
