Assay Details
Gene Symbol: Chat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009891.2 | NP_034021.1 | 7 - 8 | 942 | 68 | |
GenBank mRNA | AK046406.1 | - | 7 - 8 | 943 | 68 | |
AK047708.1 | - | 6 - 7 | 834 | 68 | ||
BC119322.1 | - | 6 - 7 | 704 | 8734071 | 68 | |
BC120589.1 | - | 6 - 7 | 704 | 40129512 | 68 | |
D12487.1 | - | 6 - 7 | 866 | 68 | ||
D12490.1 | - | 6 - 7 | 835 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
