Assay Details
Gene Symbol: Grid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008166.2 | NP_032192.2 | 13 - 14 | 2288 | 71 | |
GenBank mRNA | AK138279.1 | - | 13 - 14 | 2278 | 71 | |
D10171.1 | - | 13 - 14 | 2258 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
