Assay Details
Gene Symbol: Tbc1d14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113362.1 | NP_001106833.1 | 2 - 3 | 226 | 77 | |
GenBank mRNA | AK029986.1 | - | 2 - 3 | 227 | 77 | |
AK131724.1 | - | 2 - 3 | 259 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
