Assay Details
Gene Symbol: Tbc1d14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113362.1 | NP_001106833.1 | 3 - 4 | 962 | 106 | |
NM_133910.3 | NP_598671.3 | 2 - 3 | 781 | 106 | ||
GenBank mRNA | AB093293.1 | - | 1 - 2 | 575 | 106 | |
AK029986.1 | - | 3 - 4 | 963 | 106 | ||
BC042515.1 | - | 2 - 3 | 757 | 4020510 | 106 | |
BC086316.1 | - | 2 - 3 | 849 | 30112465 | 106 | |
BC096446.1 | - | 2 - 3 | 773 | 6307706 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
