Assay Details
Gene Symbol: Rell1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145923.4 | NP_666035.2 | 3 - 4 | 549 | 84 | |
GenBank mRNA | AK133845.1 | - | 4 - 5 | 569 | 84 | |
AK140108.1 | - | 3 - 4 | 549 | 84 | ||
AK159944.1 | - | 3 - 4 | 475 | 84 | ||
AK161061.1 | - | 3 - 4 | 475 | 84 | ||
AK182372.1 | - | 2 - 3 | 108 | 84 | ||
AK202598.1 | - | 2 - 3 | 108 | 84 | ||
BC031198.1 | - | 3 - 4 | 518 | 5059693 | 84 | |
BC066137.1 | - | 3 - 4 | 528 | 30012041 | 84 | |
BC066160.1 | - | 3 - 4 | 528 | 30012671 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
