Assay Details
Gene Symbol: Rell1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145923.4 | NP_666035.2 | 5 - 6 | 844 | 74 | |
GenBank mRNA | AK133845.1 | - | 6 - 7 | 864 | 74 | |
AK140108.1 | - | 5 - 6 | 844 | 74 | ||
AK159944.1 | - | 5 - 6 | 770 | 74 | ||
AK161061.1 | - | 5 - 6 | 770 | 74 | ||
BC023848.1 | - | 1 - 2 | 140 | 74 | ||
BC024679.1 | - | 2 - 3 | 293 | 74 | ||
BC031198.1 | - | 5 - 6 | 813 | 5059693 | 74 | |
BC066137.1 | - | 5 - 6 | 823 | 30012041 | 74 | |
BC066160.1 | - | 5 - 6 | 823 | 30012671 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
