Assay Details
Gene Symbol: Aco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007386.2 | NP_031412.2 | 2 - 3 | 402 | 95 | |
GenBank mRNA | AK027914.1 | - | 2 - 3 | 187 | 96 | |
AK085951.1 | - | 2 - 3 | 151 | 95 | ||
AK129025.1 | - | 2 - 3 | 184 | 95 | ||
AK163985.1 | - | 2 - 3 | 201 | 95 | ||
AK216262.1 | - | 1 - 2 | 112 | 95 | ||
BC005454.1 | - | 2 - 3 | 162 | 3494686 | 95 | |
BY145469.1 | - | 2 - 3 | 246 | 95 | ||
X61147.1 | - | 2 - 3 | 169 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
