Assay Details
Gene Symbol: Msh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008628.2 | NP_032654.1 | 10 - 11 | 1700 | 87 | |
GenBank mRNA | AK157842.1 | - | 10 - 11 | 1713 | 87 | |
AK166179.1 | - | 10 - 11 | 1688 | 87 | ||
BC047117.1 | - | 10 - 11 | 1708 | 4951173 | 87 | |
BC050897.1 | - | 10 - 11 | 1683 | 6401835 | 87 | |
U21011.1 | - | 10 - 11 | 1717 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
