Assay Details
Gene Symbol: Bag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145392.2 | NP_663367.1 | 2 - 3 | 276 | 64 | |
GenBank mRNA | AK083600.1 | - | 2 - 3 | 227 | 64 | |
AK144720.1 | - | 2 - 3 | 261 | 64 | ||
BC016230.1 | - | 2 - 3 | 296 | 4488760 | 64 | |
BC028944.1 | - | 2 - 3 | 259 | 5371263 | 64 | |
BY014303.1 | - | 2 - 3 | 278 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20134777 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20134779 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20134778 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P204913 | BC028944.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P204913 | AK083600.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P204913 | AK144720.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P204913 | BC016230.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P204913 | NM_145392.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P204913 | BC028944.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P204913 | AK083600.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P204913 | AK144720.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P204913 | BC016230.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P204913 | NM_145392.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 358510 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 470671 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 928817 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 5805 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 146881 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 286115 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 151363 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 279016 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 127147 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108947 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169282 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P204913 | BC028944.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P204913 | AK083600.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P204913 | AK144720.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P204913 | BC016230.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P204913 | NM_145392.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P204913 | BC028944.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P204913 | AK083600.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P204913 | AK144720.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P204913 | BC016230.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P204913 | NM_145392.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204913 | BC028944.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204913 | AK083600.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204913 | AK144720.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204913 | BC016230.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204913 | NM_145392.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204913 | BC028944.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204913 | AK083600.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204913 | AK144720.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204913 | BC016230.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204913 | NM_145392.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134765 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134801 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134774 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134764 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134816 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134804 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134806 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134778 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134779 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134768 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134783 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134784 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134759 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134812 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134777 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134785 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134797 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134809 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134763 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134766 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134803 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134807 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134790 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134761 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134817 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134771 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134792 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134818 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134814 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134799 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134781 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134794 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134810 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134791 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134773 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134787 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20134808 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04057416 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108947 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P169282 | - | - |
Whole Mouse Genome G4122A | A_51_P204913 | BC028944.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P204913 | AK083600.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P204913 | AK144720.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P204913 | BC016230.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P204913 | NM_145392.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P204913 | BC028944.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P204913 | AK083600.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P204913 | AK144720.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P204913 | BC016230.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P204913 | NM_145392.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104128 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P169282 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108947 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00371069 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00371071 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00371064 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00371066 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20134778 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20134777 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20134779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134624 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134799 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134686 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134629 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134690 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134724 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134706 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134637 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134698 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134713 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134783 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134709 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134807 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134803 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134808 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134688 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20134761 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00935929 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134748 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134773 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134794 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134732 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134785 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134791 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134763 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134774 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134804 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00936025 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134803 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134797 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134733 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134795 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134761 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00510481 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134754 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134788 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134806 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134747 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134784 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134790 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00447681 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134757 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00104809 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00104807 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00447676 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134740 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134792 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134759 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00510480 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134781 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00322889 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134808 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134764 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134779 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00510483 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134734 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134807 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134765 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134783 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00417828 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00233732 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00104804 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134743 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00447678 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134771 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20134744 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10353752 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10353753 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10353751 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1423948_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10353750 | - | - |
Mouse Genome 430 2.0 Array | 1423948_at | - | - |
Mouse Genome 430 2.0 Array | 1423948_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 213539_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 213539_PM_at | - | - |
Mouse Expression Set 430 | 1423948_PM_at | - | - |
Mouse Expression Set 430 | 1423948_at | - | - |
Murine Genome U74v2 Set | 160962_at | - | - |
Murine Genome U74v2 Set | 161129_r_at | - | - |
Murine Genome U74v2 Set | 161129_PM_r_at | - | - |
Murine Genome U74v2 Set | 160962_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1423948_at | - | - |
Mouse Genome 430A 2.0 Array | 1423948_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1423948_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1423948_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 872381 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2900324 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2900324 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002900324 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1234419 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1234419 | - | - |
mouse-6 v1.1 (Illumina) | scl16893.3_29-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2900324 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl16893.3_29-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2900324 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1234419 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1234419 | - | - |
