Assay Details
Gene Symbol: Pik3cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029094.3 | NP_083370.2 | 2 - 3 | 313 | 118 | |
GenBank mRNA | AK081350.1 | - | 2 - 3 | 508 | 118 | |
AK090116.1 | - | 2 - 3 | 190 | 118 | ||
AK154106.1 | - | 2 - 3 | 298 | 127 | ||
BC039650.1 | - | 2 - 3 | 276 | 118 | ||
CJ129150.1 | - | 2 - 3 | 314 | 118 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK090116.1, AK154106.1, BC039650.1, NM_029094.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
