Assay Details
Gene Symbol: Heph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159627.1 | NP_001153099.1 | 12 - 13 | 2355 | 62 | |
NM_001159628.1 | NP_001153100.1 | 12 - 13 | 2363 | 62 | ||
NM_010417.2 | NP_034547.2 | 12 - 13 | 2363 | 62 | ||
NM_181273.4 | NP_851790.1 | 12 - 13 | 2229 | 62 | ||
GenBank mRNA | AF082567.1 | - | 12 - 13 | 2475 | 62 | |
AK081330.1 | - | 12 - 13 | 2205 | 62 | ||
AK129194.1 | - | 12 - 13 | 2311 | 62 | ||
BC048237.1 | - | 12 - 13 | 2203 | 6474714 | 62 | |
BC054442.1 | - | 12 - 13 | 2203 | 6474713 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
