Assay Details
Gene Symbol: Wdr60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146039.3 | NP_666151.3 | 8 - 9 | 1446 | 68 | |
GenBank mRNA | AK052482.1 | - | 8 - 9 | 1410 | 68 | |
AK144138.1 | - | 8 - 9 | 1446 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
