Assay Details
Gene Symbol: Rps6ka6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025949.3 | NP_080225.2 | 21 - 22 | 2279 | 61 | |
GenBank mRNA | AK049349.1 | - | 19 - 20 | 2030 | 61 | |
BC054113.1 | - | 19 - 20 | 1952 | 30015425 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
