Assay Details
Gene Symbol: Atp7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109757.2 | NP_001103227.1 | 10 - 11 | 2555 | 121 | |
NM_009726.5 | NP_033856.3 | 10 - 11 | 2552 | 121 | ||
GenBank mRNA | AB007134.1 | - | 9 - 10 | 2391 | 121 | |
BC141395.1 | - | 10 - 11 | 2433 | 9056008 | 121 | |
U03434.1 | - | 10 - 11 | 2460 | 121 | ||
U71091.1 | - | 10 - 11 | 2456 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
