Assay Details
Gene Symbol: Atp7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109757.2 | NP_001103227.1 | 14 - 15 | 3068 | 103 | |
NM_009726.5 | NP_033856.3 | 14 - 15 | 3065 | 103 | ||
GenBank mRNA | AB007134.1 | - | 13 - 14 | 2904 | 103 | |
BC141395.1 | - | 14 - 15 | 2946 | 9056008 | 103 | |
U03434.1 | - | 14 - 15 | 2973 | 103 | ||
U03736.1 | - | 13 - 14 | 2813 | 103 | ||
U71091.1 | - | 14 - 15 | 2969 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
