Assay Details
Gene Symbol: Kif5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008448.3 | NP_032474.2 | 22 - 23 | 2860 | 65 | |
GenBank mRNA | BC090841.1 | - | 22 - 23 | 2836 | 30543821 | 65 |
U86090.1 | - | 22 - 23 | 2687 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
