Assay Details
Gene Symbol: Chpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146690.1 | NP_001140162.1 | 5 - 6 | 896 | 60 | |
NM_144807.3 | NP_659056.3 | 5 - 6 | 896 | 60 | ||
NR_027477.1 | 5 - 6 | 896 | 60 | |||
GenBank mRNA | AK007964.1 | - | 5 - 6 | 816 | 60 | |
AK032497.1 | - | 5 - 6 | 791 | 60 | ||
AY445814.1 | - | 5 - 6 | 791 | 60 | ||
BE650891.1 | - | 1 - 2 | 131 | 60 | ||
EL958096.1 | - | 4 - 5 | 496 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
