Assay Details
Gene Symbol: Atl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019717.3 | NP_062691.3 | 2 - 3 | 466 | 108 | |
GenBank mRNA | BC060501.1 | - | 2 - 3 | 371 | 5362692 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
