Assay Details
Gene Symbol: Arhgap28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172964.4 | NP_766552.3 | 9 - 10 | 1313 | 67 | |
GenBank mRNA | AK087523.1 | - | 9 - 10 | 1326 | 67 | |
AK133720.1 | - | 9 - 10 | 1155 | 67 | ||
AK141901.1 | - | 9 - 10 | 1263 | 67 | ||
AK162051.1 | - | 3 - 4 | 353 | 67 | ||
AK173147.1 | - | 8 - 9 | 940 | 67 | ||
BC066788.1 | - | 9 - 10 | 1303 | 30049271 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
