Assay Details
Gene Symbol: Abcc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033336.3 | NP_001028508.2 | 18 - 19 | 2413 | 81 | |
NM_001163675.1 | NP_001157147.1 | 18 - 19 | 2395 | 81 | ||
NM_001163676.1 | NP_001157148.1 | 17 - 18 | 2188 | 81 | ||
GenBank mRNA | AK052778.1 | - | 18 - 19 | 2396 | 81 | |
AK157723.1 | - | 18 - 19 | 2400 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
