Assay Details
Gene Symbol: Abcc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033336.3 | NP_001028508.2 | 7 - 8 | 1025 | 64 | |
NM_001163675.1 | NP_001157147.1 | 7 - 8 | 1007 | 64 | ||
NM_001163676.1 | NP_001157148.1 | 6 - 7 | 800 | 64 | ||
GenBank mRNA | AK052778.1 | - | 7 - 8 | 1008 | 64 | |
AK157723.1 | - | 7 - 8 | 1012 | 64 | ||
BC150822.1 | - | 7 - 8 | 979 | 9087733 | 64 | |
BC171974.1 | - | 6 - 7 | 754 | 9054260 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
