Assay Details
Gene Symbol: Tmtc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028651.2 | NP_082927.1 | 1 - 2 | 87 | 53 | |
GenBank mRNA | AK028333.1 | - | 1 - 2 | 68 | 53 | |
AK033947.1 | - | 1 - 2 | 88 | 53 | ||
AK036741.1 | - | 1 - 2 | 44 | 53 | ||
AK039093.1 | - | 1 - 2 | 32 | 53 | ||
AK044391.1 | - | 1 - 2 | 43 | 53 | ||
AK045271.1 | - | 1 - 2 | 49 | 53 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK028333.1, AK033947.1, AK039093.1, AK044391.1, AK045271.1, NM_028651.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
