Assay Details
Gene Symbol: Tmtc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028651.2 | NP_082927.1 | 6 - 7 | 827 | 60 | |
GenBank mRNA | AK028333.1 | - | 6 - 7 | 808 | 60 | |
AK033947.1 | - | 6 - 7 | 828 | 60 | ||
AK036741.1 | - | 6 - 7 | 784 | 60 | ||
AK039093.1 | - | 6 - 7 | 772 | 60 | ||
AK043293.1 | - | 6 - 7 | 854 | 60 | ||
AK044391.1 | - | 6 - 7 | 783 | 60 | ||
AK045271.1 | - | 6 - 7 | 789 | 60 | ||
AK049748.1 | - | 7 - 8 | 968 | 60 | ||
AK082526.1 | - | 5 - 6 | 655 | 60 | ||
BC031368.1 | - | 2 - 3 | 201 | 5029461 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
