Assay Details
Gene Symbol: Eml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114361.1 | NP_001107833.1 | 18 - 19 | 2283 | 81 | |
NM_001114362.1 | NP_001107834.1 | 16 - 17 | 2076 | 81 | ||
NM_001286567.1 | NP_001273496.1 | 18 - 19 | 2186 | 81 | ||
NM_199466.3 | NP_955760.3 | 15 - 16 | 1947 | 81 | ||
GenBank mRNA | AK139067.1 | - | 2 - 3 | 140 | 81 | |
AK144604.1 | - | 18 - 19 | 2284 | 81 | ||
AK189201.1 | - | 1 - 2 | 35 | 81 | ||
BC032258.1 | - | 4 - 5 | 431 | 81 | ||
BC065178.1 | - | 18 - 19 | 2124 | 6827397 | 81 | |
BC066099.1 | - | 15 - 16 | 1878 | 30533573 | 81 | |
BC067011.1 | - | 15 - 16 | 1896 | 6405184 | 81 | |
BC070427.1 | - | 16 - 17 | 2016 | 6413041 | 81 | |
BC079619.1 | - | 15 - 16 | 1878 | 30533573 | 81 | |
DQ230326.1 | - | 17 - 18 | 1966 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
