Assay Details
Gene Symbol: Plekhh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177606.4 | NP_808274.2 | 18 - 19 | 2904 | 128 | |
GenBank mRNA | AK029252.1 | - | 18 - 19 | 2906 | 128 | |
AK173337.1 | - | 15 - 16 | 2559 | 128 | ||
BC125583.1 | - | 18 - 19 | 2869 | 40130098 | 128 | |
BC137804.1 | - | 18 - 19 | 2866 | 8860823 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
