Assay Details
Gene Symbol: Plekhh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177606.4 | NP_808274.2 | 25 - 26 | 3869 | 90 | |
GenBank mRNA | AK029252.1 | - | 25 - 26 | 3871 | 90 | |
AK086794.1 | - | 5 - 6 | 699 | 90 | ||
AK173337.1 | - | 22 - 23 | 3524 | 90 | ||
BC125583.1 | - | 25 - 26 | 3834 | 40130098 | 90 | |
BC137804.1 | - | 25 - 26 | 3831 | 8860823 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
