Assay Details
Gene Symbol: Rmdn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201361.2 | NP_958749.1 | 4 - 5 | 828 | 101 | |
GenBank mRNA | AK034617.1 | - | 4 - 5 | 830 | 101 | |
AK139382.1 | - | 4 - 5 | 1047 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
