Assay Details
Gene Symbol: Eif5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198303.2 | NP_938045.2 | 11 - 12 | 2214 | 66 | |
GenBank mRNA | AK145146.1 | - | 11 - 12 | 2101 | 66 | |
BC103806.1 | - | 10 - 11 | 2967 | 66 | ||
CN670500.1 | - | 2 - 3 | 247 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
