Assay Details
Gene Symbol: Lrp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008513.3 | NP_032539.2 | 19 - 20 | 4226 | 70 | |
GenBank mRNA | AF064984.1 | - | 19 - 20 | 4216 | 70 | |
AF077847.1 | - | 19 - 20 | 4104 | 70 | ||
AK090382.1 | - | 10 - 11 | 1901 | 70 | ||
AK147068.1 | - | 19 - 20 | 4219 | 70 | ||
AK147231.1 | - | 19 - 20 | 7966 | 70 | ||
AK147330.1 | - | 19 - 20 | 7954 | 70 | ||
AK220186.1 | - | 19 - 20 | 4179 | 70 | ||
BC011374.1 | - | 19 - 20 | 4208 | 3154246 | 70 | |
BC094610.1 | - | 19 - 20 | 4213 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
