Assay Details
Gene Symbol: Sgk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037759.1 | NP_001032848.1 | 2 - 3 | 325 | 101 | |
NM_133220.2 | NP_573483.1 | 3 - 4 | 444 | 101 | ||
NM_177547.3 | NP_808215.2 | 2 - 3 | 337 | 101 | ||
GenBank mRNA | AF312007.1 | - | 3 - 4 | 386 | 101 | |
AK028722.1 | - | 3 - 4 | 389 | 101 | ||
AK030314.1 | - | 2 - 3 | 326 | 101 | ||
AK031133.1 | - | 3 - 4 | 425 | 101 | ||
AK031328.1 | - | 2 - 3 | 266 | 101 | ||
AK143836.1 | - | 3 - 4 | 432 | 101 | ||
AK146561.1 | - | 2 - 3 | 280 | 101 | ||
AK151181.1 | - | 3 - 4 | 389 | 101 | ||
AK151793.1 | - | 2 - 3 | 317 | 101 | ||
AK151867.1 | - | 3 - 4 | 421 | 101 | ||
AK166414.1 | - | 2 - 3 | 266 | 101 | ||
AK166531.1 | - | 2 - 3 | 287 | 101 | ||
AK171120.1 | - | 2 - 3 | 313 | 101 | ||
AK171186.1 | - | 2 - 3 | 313 | 101 | ||
AK171678.1 | - | 2 - 3 | 296 | 101 | ||
AK171959.1 | - | 2 - 3 | 374 | 101 | ||
AK172323.1 | - | 2 - 3 | 287 | 101 | ||
BB856423.1 | - | 2 - 3 | 361 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
