Assay Details
Gene Symbol: Gabra6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099641.2 | NP_001093111.1 | 1 - 2 | 346 | 67 | |
NM_008068.3 | NP_032094.2 | 1 - 2 | 346 | 67 | ||
GenBank mRNA | AF256197.1 | - | 1 - 2 | 42 | 67 | |
AF256198.1 | - | 1 - 2 | 42 | 67 | ||
AK135145.1 | - | 1 - 2 | 326 | 67 | ||
AK137331.1 | - | 1 - 2 | 326 | 67 | ||
BC145158.1 | - | 1 - 2 | 64 | 9053709 | 67 | |
BC145702.1 | - | 1 - 2 | 64 | 40131005 | 67 | |
X51986.1 | - | 1 - 2 | 282 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
