Assay Details
Gene Symbol: Arhgap12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039692.1 | NP_001034781.1 | 7 - 8 | 1695 | 70 | |
GenBank mRNA | AK031648.1 | - | 7 - 8 | 1539 | 70 | |
AK077063.1 | - | 7 - 8 | 1696 | 70 | ||
BC024633.1 | - | 1 - 2 | 92 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC024633.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
