Assay Details
Gene Symbol: Usp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011678.2 | NP_035808.2 | 9 - 10 | 1214 | 88 | |
GenBank mRNA | AK143582.1 | - | 9 - 10 | 1149 | 88 | |
AK149964.1 | - | 9 - 10 | 1172 | 88 | ||
AK169372.1 | - | 9 - 10 | 1193 | 88 | ||
AK169933.1 | - | 9 - 10 | 1184 | 88 | ||
AK171271.1 | - | 9 - 10 | 1184 | 88 | ||
AK171633.1 | - | 9 - 10 | 1167 | 88 | ||
AK220512.1 | - | 9 - 10 | 1171 | 88 | ||
BC011341.1 | - | 8 - 9 | 991 | 3661165 | 88 | |
BC066180.1 | - | 9 - 10 | 1124 | 88 | ||
BC066865.1 | - | 9 - 10 | 1124 | 88 | ||
L00681.1 | - | 9 - 10 | 1177 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
