Assay Details
Gene Symbol: Taf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290729.1 | NP_001277658.1 | 24 - 25 | 3935 | 118 | |
GenBank mRNA | AB299229.1 | - | 24 - 25 | 3862 | 118 | |
AF081117.1 | - | 2 - 3 | 255 | 118 | ||
AK046668.1 | - | 3 - 4 | 424 | 118 | ||
BC047418.1 | - | 14 - 15 | 2048 | 118 | ||
BC094568.1 | - | 6 - 7 | 1219 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
